SST Series Heat-Tolerant Brushes, Insulative, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Nylon bristle applicator brushes with a lightweight stainless steel handle.
- Nylon bristle material is FDA approved
- Stainless steel handle is chemical resistant and non-magnetic
- Maximum use temperature of 250°F for short term exposures and 200°F for continuous or long term exposures
Manufactured using strong mechanical clamping for maximum fiber retention, and is adhesive-free to prevent contamination on surface areas caused by leaching.
Customers Who Bought This Also Bought