Wood Scratch Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Antistatic brushes are ideal for cleaning circuit boards and SMT assemblies.
- Plywood handles
- Different versions available
One row brush has extra-fine, brass-fill fibers hand laced for soft cleaning, maximum retention, and durability.
The three row brush is small and economical with “scratch” or “toothbrush” style bristles.
The larger sratch brush has four rows of bristles.
Customers Who Bought This Also Bought