Cryogenic Vial and Vaccine Vial Grippers
Supplier: Heathrow Scientific
New Product
Certificates
About this item
The perfect grip for optimal precision.
- Durable grippers are made of reinforced polypropylene, which is cold-tolerant and resistant to everyday wear
- The designated squeeze target ensures maximum precision and grip with every use
- Each style is available in a pack of 5 assorted colors for easy identification and organization
- A combo pack with one of each style is also available
These two styles of hand-operated grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively.
Customers Who Bought This Also Bought