Test Tube Brushes, Tufted End, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Test tube brushes are designed for cleaning test tubes and narrow mouth bottles.
- White natural bristles with black nylon tuft
- Twisted wire handle with loop for hanging
Customers Who Bought This Also Bought