Narrow Tubing Brushes, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Feature short, close bristles for effective cleaning of narrow tubing, burets, pipets, and funnel stems.
Natural white bristles on twisted wire handle with a convenient loop for hanging.
Customers Who Bought This Also Bought