Centrifuge Tube Brushes, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Tapered natural bristle construction for cleaning centrifuge tubes, vials, and small funnels.
- Flexible twisted wire handle has a convenient hanging loop
- Select models available with black nylon tufted ends
- Fan tip model provides effective cleaning for conical tubes
Customers Who Bought This Also Bought