Beaker and Jar Brushes, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Designed for heavy-duty cleaning of beakers, jars, and other wide-mouth glassware.
- Heavy tufting at the tip reduces scratching and improves cleaning capability
- Handle has an opening for hanging
Long-lasting bristles are spirally wound into grooves in the smooth wood handle.
Customers Who Bought This Also Bought