Nalgene® Rectangular Carboys with Spigot and Handle, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
For dispensing solutions, media, and sterile water.
- 8.9 cm (3¹/₂") wide mouth for easy filling and cleaning
- Sturdy handles
- Graduated in liters and gallons
- Leakproof
- Autoclavable
Note: Autoclave only when empty, using a properly vented closure. Dismount and disassemble spigot before autoclaving.
Ordering information: Supplied with carboy, polypropylene spigot, and 100-415 screw closure. For replacement caps, see cap 16219-106 in 16219-030 series.
Frequently Bought Together