Rat Recombinant MSP1E3D1-His (from E. coli)
Fournisseur: CUBE BIOTECH
New Product
Certificats
A propos de cet article
Membrane scaffold protein, rat origin.
- Available origins: Human, mouse, rat
- Extinction coefficient (in water) ε280: 29400 M⁻¹cm⁻¹ (Human origin); 32430 M⁻¹cm⁻¹ (Mouse origin); 36440 M⁻¹cm⁻¹(Rat origin)
- Buffer: 20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: N-Terminal His-tag
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences for the stabilization of membrane proteins in a lipid environment after the removal of the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1E3D1 (His-tagged) creates nanodiscs of 12 to 14 nm in diameter. This product contains a His-tag, keep that in mind when choosing the affinity tag for your membrane protein. This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.
Customers Who Bought This Also Bought