Mouse Recombinant MSP1D1 (from E. coli)
Fournisseur: CUBE BIOTECH
New Product
Certificats
A propos de cet article
Membrane scaffold protein, mouse origin.
- Extinction coefficient (in water) ε280: 25440 M⁻¹cm⁻¹ (Human origin), 20970 M⁻¹cm⁻¹ (Mouse origin)
- Buffer:20 mM TRIS pH 7.4; 100 mM NaCl; 0.5 mM EDTA
- Affinity tag: none
Membrane scaffold proteins derive from apolipoprotein (apo; A-I). They are used in membrane protein sciences to stabilize membrane proteins in a lipid environment after removing the cell membrane. The size of the nanodisc is determined by the MSP that was used to create the disc. MSP1D1 (His-tagged) creates nanodiscs of 9 to 10 nm in diameter.
This product is available with different species as its background. This is important for e.g. for secondary antibodies of a western blot.
Customers Who Bought This Also Bought