Vinyl-Coated Lead Rings 'C' Shape, Blue
Catalog # 77395-448
Supplier: Heathrow Scientific
New Product
Specifications
- Description:Vinyl-coated lead rings ('C' shape), blue
- Cat. no.:77395-448
Specifications
About this item
Stabilize vessels and labware on the bench or in water baths.
- Vinyl-coated lead ring will not mark or scratch benchtops and labware
- C-shape style allows some manual size variation for a customized fit
- Fits 1000 to 4000 ml
The lead ring is perfect for stabilizing vessels and labware or for water bath immersions.
Customers Who Bought This Also Bought