ContactUsLinkComponent
Ice Pan Cool Container, 4 L, Blue
Ice Pan Cool Container, 4 L, Blue
Catalog # 77395-440
New Product
CONTAINER COOL 4L ICE PAN BLUE
Ice Pan Cool Container, 4 L, Blue
Catalog # 77395-440
New Product
Specifications
  • Capacity:
    4 L
  • Color:
    Blue
  • Description:
    Ice pan cool container
  • Cat. no.:
    77395-440
Specifications
About this item
Durable containers for chilling temperature-sensitive samples.
  • Suitable for low temperature use.; ideal for ice, ice slurries and CO₂ pellets
  • Made from polyurethane with metal inserts for stability
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Tapered design for easy stacking
  • Precise pour spout and large handles
  • Lid nests under bucket for convenience and to save space
  • Large handles for safer transportation
Customers Who Bought This Also Bought