ContactUsLinkComponent
Clearly Safe® Acrylic Safety Spectacles Dispenser Counter or Wall Mount, Blue
Clearly Safe® Acrylic Safety Spectacles Dispenser Counter or Wall Mount, Blue
Catalog # 77395-324
New Product
DISPENSER ACRYLIC 20EYEGLASSES BLUE
Clearly Safe® Acrylic Safety Spectacles Dispenser Counter or Wall Mount, Blue
Catalog # 77395-324
New Product
Specifications
  • Color:
    Blue
  • Description:
    Acrylic safety spectacles dispenser
  • Cat. no.:
    77395-324
Specifications
About this item
Ergonomically angled for easy dispensing.
  • Translucent acrylic dispensers provide easy access to safety accessories
  • Hinged lid protects glasses from dust and debris
  • Set on counter or wall mount (screws included)
  • Large dispenser holds approx. 20 glasses, small approx. 10 glasses
  • Great for holding other personal items
Customers Who Bought This Also Bought