Order Entry
ContactUsLinkComponent
Anti-Strep Antibody (HRP (Horseradish Peroxidase))
Anti-Strep Antibody (HRP (Horseradish Peroxidase))
# de catalogue 77393-152
Fournisseur:  CUBE BIOTECH
New Product
STREP ANTIBODY HRP
Anti-Strep Antibody (HRP (Horseradish Peroxidase))
# de catalogue 77393-152
Fournisseur:  CUBE BIOTECH
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Conjugation:
    HRP (Horseradish Peroxidase)
  • Size:
    100 µg
  • Cat. no.:
    77393-152

Spécifications

A propos de cet article

This is an antibody specifically for targeting a Strep-(WSHPQFEK)-tag on a protein.

  • Antibody name: Strep antibody HRP
  • Cell line/origin: Mouse purified and conjugated to horseradish peroxidase
  • Subclass: IgG
  • Form: Suspension in PBS containing 5 mg/ml bovine serum albumin; approx. 0.05% ProClin 300 and stabilizing agents
  • Specificity: Strep-tag® II; the antibody was produced in mouse using Strep-tag® II peptide conjugated to keyhole limpet hemocyanin as immunogen
  • Detections: N- or C-terminal Strep-tag® II and Twin-Streptag®
  • Dilutions: Dilute 1:10000 or higher for detection of Streptag® II fusion proteins coated to microplates. Dilute 1:4000 for chromogenic and 1:32000 for chemiluminescence detection of Strep-tag® fusion proteins after Western blotting
  • Amount: 75 µl
  • Shipment temperature: Room tempareature
  • Storage: Recommended: 2 to 8 °C

It can be used in Western Blots and ELISAs.Its origin is mouse and should be stored at −20 °C for the long term.

Type: Recombinant
Antigen: Strep Antibody
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host:
Isotype:
Reactivity

Customers Who Bought This Also Bought