Order Entry
ContactUsLinkComponent
Anti-Strep Antibody
Anti-Strep Antibody
# de catalogue 77393-150
Fournisseur:  CUBE BIOTECH
New Product
STREP ANTIBODY 100 UG
Anti-Strep Antibody
# de catalogue 77393-150
Fournisseur:  CUBE BIOTECH
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    100 µg
  • Shipping temperature:
    Room temperature
  • Cat. no.:
    77393-150

Spécifications

A propos de cet article

This is an antibody specifically for targeting a Strep-(WSHPQFEK)-tag on a protein.

  • Antibody name: Strep antibody
  • Cell line/origin: Mouse Hybridoma
  • Produced using Strep-tag II peptide conjugated to keyhole limpet hemocyanin as immunogen
  • Subclass: IgG
  • Suitable for: Western blot, ELISA, FACS, Immune fluorescence
  • Form: 100 µg lyophilized from NH₄CO₃, contains 10 mg sucrose per mg protein
  • Reconstitution of lyophilized antibody: Reconstitute antibody in PBS buffer at concentrations between 0.2 and 1.0 mg/ml (i.e. dissolve with 0.5 to 0.1 PBS)
  • Recommended dilution for Western Blot, chromogenic detection: 0.2 to 0.5 µg/ml antibody diluted in PBS containing 2% BSA. Use rabbit anti-mouse conjugated to peroxidase as secondary antibody
  • Recommended dilution for Western Blot, chemiluminescence detection: 20 ng/ml antibody diluted in PBS
  • Recommended dilution for FACS: 10 µg/ml diluted in PBS, using e.g. goat anti mouse IgG AB conjugated to Cy2 (GE Healthcare, 1:400 dilution)
  • Recommended dilution for IF: 10 to 5 µg/ml antibody diluted in PBS (use secondary anti-mouse antibody)
  • Shipment temperature: Room tempareature
  • Storage: Store as lyophilized powder for up to 12 months at 2 to 8 °C. Alternatively, alinquot contents and freeze at −20 °C for up to 6 months. Avoid repeated freeze-that cycles

The Strep antibody can be used in Western Blots and ELISAs. Its origin is mouse and should be stored at −20 °C for the long term.

Type: Recombinant
Antigen: Strep Antibody
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity

Customers Who Bought This Also Bought