Order Entry
ContactUsLinkComponent
Anti-GST Antibody
Anti-GST Antibody
# de catalogue 77393-148
Fournisseur:  CUBE BIOTECH
New Product
GST ANTIBODY 100 UG
Anti-GST Antibody
# de catalogue 77393-148
Fournisseur:  CUBE BIOTECH
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Size:
    100 µg
  • Shipping temperature:
    Shipped at ambient temperature
  • Cat. no.:
    77393-148

Spécifications

A propos de cet article

This is an antibody specifically for targeting a GST-tag on a protein.

  • Cell line: Mouse, Hybridoma
  • Suitable for: Western blot, ELISA
  • Detection limit Western blot, chromogenic detection: Not determined
  • Detection limit Western blot, chemiluminescent detection: 500 pg
  • Quantity: 100 µg
  • Physical state: Dried lyophilized powder
  • Reconstitution of lyophilized antibody: Add 500 µl ddH₂O to obtain a solution of 0.2 mg/ml concentration
  • Recommended working dilution: 1:1000
  • Shipment temperature: Shipped at room temperature (RT)
  • Short-term storage: After resuspension, store vial at 2 to 8 °C for up to 3 months. Centrifuge product if not clear after standing at room temperature and/or heating to 37 °C
  • Long-term storage: Store as lyophilized powder for up to 12 months at 2 to 8 °C. Alternatively, alinqout contents and freeze at −20 °C for up to 6 months. Avoid repeated freeze-thaws

The GST antibody can be used in Western Blots and ELISAs. Its origin is mouse and should be stored at −20 °C for the long term.

Type: Recombinant
Antigen: GST Antibody
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity

Customers Who Bought This Also Bought