To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Size:3×10 ml
- Cat. no.:77393-122
Spécifications
A propos de cet article
The PureCube HIC starter set includes 10 ml of each of the following HIC resins: PureCube butyl agarose, PureCube phenyl agarose, and PureCube octyl agarose.
- Usage: As a matrix for Hydrophobic Interaction Chromatography (HIC)
- Specificity: Hydrophobic biomolecules and proteins
- Bead size: 40 μm
- Bead Ligand: Octyl groups
- Octyl group density: 40 µmol/ml
- BSA binding capacity: 10 mg/ml
- lysozyme binding capacity: 20 mg/ml
- pH stability: 2 to 13
- Filling quantity: Delivered as a 50% suspension
- Matrix: 7.5% cross-linked agarose
- Short term storage: In neutral buffer at 4 °C
- Long term storage: 20 mM sodium acetate, 20% ethanol, pH 6.5 at 4 °C
- Chemical structure of Octyl ligand: Octyl group structure
Hydrophobic Interaction Chromatography (HIC) is a reliable method to separate proteins and other biomolecules based on their hydrophobicity. Our PureCube HIC starter set provides newcomers to the HIC method an excellent place to start. Alternatively, it can be used for seasoned users of HIC as a collection of resins to screen through when working with a new protein or biomolecule.