Nalgene® Air-Tight Pails, Low-Density Polyethylene, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Close-fitting, airtight covers make these pails useful for specimens, storage and general utility
LDPE construction is suitable for use with formalin. Handle is a nylon-coated wire bail. Lids are white.
Certifications: Meets OSHA Standard 29 CFR 1910.1030 for use as protection against bloodborne pathogens.
Ordering information: Comes with lids and handles.
Customers Who Bought This Also Bought