Nalgene® Rectangular Carboys with Spigot and Handle, Polycarbonate, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Transparent carboy features molded-in graduations
- Leakproof
For replacement caps, see cap 16219-106 in 16219-030 series.
Note: Before autoclaving, set cap on top of carboy without engaging threads and disassemble spigot.
High-impact strength enables reliable cold room use. Wide mouth allows easy, efficient cleaning. Autoclave only when empty. Cap size: 100-415.
Ordering information: Supplied with carboy, polypropylene screw cap, and spigot.
Customers Who Bought This Also Bought