Test Tube Brushes, Radial Tip, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Test tube brushes are designed for cleaning test tubes and narrow mouth glassware.
- Rounded tuft at tip facilitates cleaning and protects glassware
- Available with white nylon or white natural bristles
- Twisted wire handle
Customers Who Bought This Also Bought