Beaker, Jar, and Cylinder Brushes, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Completely rounded tip provides safe, effective cleaning for wide mouth glassware such as beakers, jars, and cylinders.
- Extra dense natural black bristles are mounted on twisted wires into a durable wood handle
- Double-tuft extensions at the tip for reaching into corners
- Available with two or four rows of bristles
Customers Who Bought This Also Bought