Barnstead/Thermolyne Locator® Cryobiological Storage System Boxes, Thermo Scientific
Catalog # 55709-903
Supplier: Thermo Fisher Scientific
Specifications
- No. of Holes:25
- Description:For Locator 4
- Dimensions:76×76 mm (3×3")
- Cat. no.:55709-903
- Supplier No.:CS509X3
Specifications
About this item
Nalgene® Cryoplastic boxes are of durable, ultra-low temperature polycarbonate construction
One-way lid reduces the chances of sample spillage. A printed grid system on the lid facilitates easy logging and retrieval of samples.
Customers Who Bought This Also Bought