Reagent Reservoir Chillers, 50/55 ml
Fournisseur: Heathrow Scientific
New Product
Certificats
A propos de cet article
Ice-free, non-toxic method to extend the working period and preserve integrity for temperature-sensitive reagents.
- Reagents are kept at less than 5 °C (41 °F) for approximately 1 hour
- Eliminates the risk of contamination from messy ice baths
- Not for use with liquid nitrogen
- Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
- Stackable design saves storage and work space
- 2-year warranty
Engineered custom fit design for Heathrow Scientific 50 ml or 100 ml reservoirs to ensure a maximum contact for longer uniform cooling.
Customers Who Bought This Also Bought