SST Series Antistatic Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
All SST brushes feature strong mechanical fiber clamping for maximum fiber retention and no adhesive to leach out and contaminate surface areas
Stainless steel handles. Four sizes to fit most benchwork needs.
Customers Who Bought This Also Bought