Applicator Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
This applicator brush is an inexpensive utility duster/cleaner that is an excellent replacement for disposable acid brushes that quickly shed their fibers.
- Adhesive-free, to prevent leaching and contamination of surface areas
- Anti-static
Brush fibers are secured using a zinc plated steel handle, substituting adhesives with 5 tons of mechanical clamping.
Customers Who Bought This Also Bought