Counter Duster Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
All fill materials on these duster brushes are staple set into a clear, lacquered hardwood handle with a hang-up hole
For fine dusting, choose the horsehair (22940-760) or flagged plastic (22940-762) bristle brush.
For medium dusting, use the black tampico brush (22940-764).
The heavy-duty mill duster (22940-766) with brown plastic bristles has no side flare and a heavily filled front for aggressive dusting.
Brushes feature 5 rows of bristles widthwise and 15 rows of bristles lengthwise.
Customers Who Bought This Also Bought