AB Series Acid Brushes, Gordon Brush
Fournisseur: Gordon Brush
New Product
Certificats
A propos de cet article
These disposable acid brushes with horsehair bristle and a rolled tin handle are ideal for applying acid flux, glue, lubrication, and other coatings or using for small-area cleaning and the removal of debris.
- Horsehair fill material is crimped-set at one end within a rolled tin handle
- Throw-away acid brushes: May lose fibers under repeated use
These horsehair fill acid brushes can be used in an ESD sensitive environment providing the brush remains saturated in liquid when being used.
Customers Who Bought This Also Bought