Instrument Cleaning Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Instrument cleaning brushes with nylon or stainless steel bristles. Designed for cleaning small areas, including instruments and pinpoint cleaning.
- Small tight head is mechanically clamped into a stainless steel tube and inserted into an acetal or static dissipative acetal handle
- Available straight or angled
- Heat tolerant
Customers Who Bought This Also Bought