Heat-Tolerant/Cleanroom Applicator Brushes, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Pencil shaped applicator brushes with rigid acetal handle and Nylon bristles.
- Can withstand most sterilizing environments
- Bristles are epoxy set with high temperature adhesive that is autoclavable
Brushes must be cleaned prior to cleanroom use.
The brush is typically used in situations where a round brush is needed to clean fine areas, and for applying liquids or fluids in a high temperature setting.
Customers Who Bought This Also Bought