Rotator Genie™ Rotators, 230 V
Fournisseur: Scientific Industries
Certificats
A propos de cet article
This Rotator is a compact but rugged lab rotator available in two interchangeable platform configurations providing the versatility to hold virtually any kind of container.
- 230 V model
- Rugged construction
- Interchangeable configurations
- Holds any kind of container
Choose from a Standard Platform with Retaining Bands model (no additional accessories necessary), or a Stainless Steel Magnetic Platform model with clip plate system (clip plates sold separately). Uniquely designed for a wide selection of motions from gentle rolling to forceful end over end mixing. Ideal for Cold Rooms and Incubators. (Will easily fit in most incubators).
Certifications: CE marked.
Ordering information: Standard Platform with Retaining Bands model does not require additional accessories. Magnetic Platform model requires clip plates sold separately. Platforms are interchangeable. The 230 V models are supplied without a plug.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89202-302 and 89202-306) are also available, please see separate product page.
Spécifications
- Catalog No:
- 89202-304
- 89202-308
- Overall dimensions:
- 400W x 175D (290 platform horizontal) x 400H mm Overall (15.8 x 7.0 (11.5 platform horizontal) x 15.8 in) 260 x 260mm (101/4 x 101/4") Standard Platform
- 400W x 175D (290 platform horizontal) x 400H mm Overall (15.8 x 7.0 (11.5 platform horizontal) x 15.8") 290 x 315mm ( 11.5 x 12.5") Magnetic Platform
- Platform dimensions:
- 260W x 260H mm (10.25 x 10.25") Standard Platform; 290W x 315H mm (11.5 x 12.5") Magnetic Platform
- 260W x 260H mm (10.25 x 10.25") Standard Platform; 290W x 315H mm (11.5 x 12.5") Magnetic Platform
- Weight:
- 5.2 Kg (111/2 lbs.)
- 5.2 Kg (111/2 lbs.)