KIMBLE® KIMAX® Reservoir Bottles with Bottom Hose Outlet, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Reservoir bottles feature a narrow mouth, tooled top, and white enamel marking spot.
Glass hose connection outlet is fused to the bottle. Sizes 250 and 500 ml accept 7.9mm (⁵/₁₆") I.D. flexible tubing; 1000 ml and larger sizes accept 9.5 mm (³/₈") Int. Ø tubing.
Certifications: Borosilicate glass conforms to ASTM E438, Type I, Class A.
Customers Who Bought This Also Bought