ContactUsLinkComponent
Universal Pipette Wall Mount
Universal Pipette Wall Mount
Catalog # 77395-242
New Product
WALL MOUNT HOLDER PIPETTE UNIVERSAL PK2
Universal Pipette Wall Mount
Catalog # 77395-242
New Product
Specifications
  • Color:
    Assorted; blue, green, purple, yellow, and clear
  • Description:
    Four vertical mount holders
  • Cat. no.:
    77395-242
Specifications
About this item
Wall mounted storage solution.
  • Secure and adaptable
  • ABS material
The Universal pipette holder wall-mounted storage solution saves valuable benchtop space by holding and storing pipettes. The pipette holder comes with two wall mounting brackets that can hold two pipettes each and is supplied with different colored upper and lower segments, that easily slide into place. This enables labs to color-code their pipette holders for quick identification purposes, or a user can personalize the holder in the colors of their choice.
Once mounted, the pipette holder can securely hold virtually every known brand of pipette. One handed removal or replacement of pipettes is made easy with the segments unique design that includes traction grips that help center and hold the pipettes in a vertical position which helps eliminate potential contamination.
Customers Who Bought This Also Bought