PYREX® Aspirator Bottles, with Tubing Outlet, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PYREX®, borosilicate glass, without stoppers.
- Excellent chemical resistance
- High thermal resistance, autoclavable
- Minimal thermal expansion giving relatively high resistance to temperature changes
Bottles with tubular sidearm outlet facilitates attachment of flexible tubing and it withstands repeated wet or dry autoclaving. It is designed for use as a delivery or storage container for solutions.
Tubular sidearm outlet facilitates attachment of flexible tubing. Withstands repeated wet or dry autoclaving.
Spécifications
- Catalog No:
- 16331-370
- 16335-100
- 16331-372
- 89090-728
- 89090-702
- 89091-400
- 89090-834
- 89091-240
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey