Micro-Porous Boiling Chips, Walter Stern
# de catalogue 26397-409
Fournisseur: Walter Stern
Spécifications
- Description:Micro-porous boiling chips
- Cat. no.:26397-409
- Package:227 g
- Supplier No.:501A
Spécifications
A propos de cet article
Treated, micro-porous, chemically inert carbon chips eliminate “bumping” in either aqueous or non-aqueous boiling liquids.
- Maintain smooth boiling, either at atmospheric pressure or under vacuum
- Chips also prevent oxidation or decomposition
Particularly recommended for fractional distillations, evaporations, extractions, refluxings, condensations, and other boiling operations.
Packaging: Supplied in a 226.8 g (8 oz.) jar.
Customers Who Bought This Also Bought