ContactUsLinkComponent
Orbital Shakers, 230 V, KS 260
Orbital Shakers, 230 V, KS 260
Fournisseur:  IKA Works
Certificats
Orbital Shakers, 230 V, KS 260
Orbital Shakers, 230 V, KS 260
Fournisseur:  IKA Works
Certificats
Spécifications
  • Catalog No:
  • 33998-358
  • 33998-362
  • Max. load capacity:
  • 7.5 kg (16.5 lbs.)
  • 7.5 kg (16.5 lbs.)
  • Weight:
  • 8.1 kg (17.9 lbs.)
  • 8.1 kg (17.9 lbs.)
  • Dimensions:
  • 360W x 98D x 420H mm (143/8 x 37/8 x 161/2")
  • 360W x 98D x 420H mm (143/8 x 37/8 x 161/2")
  • Operating temperature range:
  • 5...50 °C (41...122 °F); RH <=80%
  • 5...50 °C (41...122 °F); RH <=80%