Pearl® Adjustable Micro Pipettes, 12-Channel
Fournisseur: Heathrow Scientific
New Product
Certificats
A propos de cet article
Precision liquid handling.
- Each pipette is calibrated and a calibration certificate is supplied
- Internal metal parts for increased accuracy and calibration stability
- Separate tip ejector with the exception of the 500 ml (120105)
- Large digit volume display
- Ergonomic, slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
- 3-position volume display
- Volume adjustment and low-force dispensing all in one button for convenience
- Finger hook
- Volume locking ring prevents accidental volume changes
- Color coded and labeled with volume range (both top and side colored components for quick visual confirmation)
- Precision metal components for both shaft and barrel ensure reliable performance and long life
- ABS material
The Pearl® is an twelve-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires ²/₃ the operation pressure of most standard pipettes.
Certifications: Manufactured in accordance with ISO 8655 and GMP regulations.
Ordering information: A complete range of single- and multi-channel pipette volumes to cover all your possible needs is available. Contact your Avantor representative for more information.
Customers Who Bought This Also Bought