Camel Hair Brushes, Mill-Rose
Fournisseur: Mill-Rose
Certificats
A propos de cet article
Soft tapered brushes, originally designed for the fine arts, are excellent for dusting balances and delicate laboratory equipment.
Natural, washable camel hair is attached to hardwood handles by metal ferrules.
Customers Who Bought This Also Bought