VWR® Long Spatula, Porcelain
# de catalogue 89038-200
Fournisseur: VWR International
Spécifications
- Length:20.3 cm (8")
- Cat. no.:89038-200
Spécifications
A propos de cet article
Chemical-porcelain tool features a spatula on one end and knob on the other.
- High chemical resistance
- Chemical-porcelain construction
- Glazed for added protection
Customers Who Bought This Also Bought