Tube Storage Box, 81-Well
Supplier: Heathrow Scientific
New Product
Certificates
About this item
Holds 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials.
- Microtube storage box
- Hold 1.5 and 2.0 ml microcentrifuge tubes and cryogenic vials
- Stylish and identifiable
- Made of translucent, vibrantly colored polypropylene allows for visibility inside box and color coding identification
- Tube capacity is indicated on the outside the box within a distinctive wave design
- Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area
- Robust
- Easy open and secure lid fitted with either friction fit or snap clasp designs
- Stackable, saving valuable space in the lab
The 81-well microtube storage box has a compact footprint for microtube and cryogenic vial storage.
Customers Who Bought This Also Bought