Niptec 1000 Series Cutters, Aven Tools
Fournisseur: Aven Tools
Certificats
A propos de cet article
Niptec 1000 Series lightweight cutters are designed with ergonomically designed ESD safe grips provide excellent operator comfort. Due to the small size of the cutting blades, which are angled at 21°, this compact tool can produce a very fine, flush cut.
- Highly durable and long-lasting cutters
- Narrow cutting head for access into dense areas
Hardened cutting blades for consistent cutting performance. Smooth cutting action with less force due to the specially designed return spring.
Customers Who Bought This Also Bought