VWR® Spatulas, PTFE-Coated with Wooden Handles
Supplier: VWR International
Certificates
About this item
Sturdy, economical tools for handling, transferring, blending, or collecting chemical and biological samples.
- Economical
- Sturdy construction
- Non-sticking and non-corrosive
Spatulas feature a PTFE-coated stainless steel blade and a wooden handle. They can be used to transfer sticky or corrosive materials without scratching or damaging container surfaces. Multiple sizes allow for varied laboratory applications.
Customers Who Bought This Also Bought