VWR® Flat/Spoon Spatulas, Stainless Steel
# de catalogue 82027-532
Fournisseur: VWR International
Spécifications
- Length:22.9 cm (9")
- Cat. no.:82027-532
Spécifications
A propos de cet article
Ideal for use in the transfer, collection, and handling of chemical or biological samples.
- Stainless steel construction
- Rustproof
- Nonstick surface
These rustproof stainless steel utensils feature a flat spatula at one end and a spoon at the other. Substances will not stick to spatula surfaces.
Customers Who Bought This Also Bought