ContactUsLinkComponent
Orbital Shakers, 230 V, KS 130
Orbital Shakers, 230 V, KS 130
Fournisseur:  IKA Works
Certificats
Orbital Shakers, 230 V, KS 130
Orbital Shakers, 230 V, KS 130
Fournisseur:  IKA Works
Certificats
Spécifications
  • Catalog No:
  • 33998-350
  • 33998-354
  • Max. load:
  • 2 kg (4.4 lbs.)
  • 2 kg (4.4 lbs.)
  • Dimensions:
  • 27W x 31.6D x 9.8H cm (105/8 x 127/16 x 37/8")
  • 27W x 31.6D x 9.8H cm (105/8 x 127/16 x 37/8")
  • Operating temperature range:
  • 5...40 °C (41...104 °F)
  • 5...40 °C (41...104 °F)