Multi-MicroPlate Genie® Mixer Shaker, 230 V
# de catalogue 89202-336
Fournisseur: Scientific Industries
Irregular Voltage
Spécifications
- Max. capacity:4 microplate, any type with standard SBS footprint
- Overall dimensions:335D x 240W x 165H mm (131/5 x 92/5 x 61/2")
- Orbit:1.0 mm
- Weight:3.7 Kg (8 lbs.)
- Pulse ON/OFF:0.1 seconds - 99 minutes (digital models only)
- Cat. no.:89202-336
- Electrical:230 V
- Speed:750-2850
- Timer:0 - 99 min or continuous
- Supplier No.:SI-4001A
Spécifications
A propos de cet article
This mixer available in analog or digital models holds four microplates, and has an aggressive speed for increased mixing efficiency in every well
- 230 V model
- Analog or digital
- Aggressive speed
- Small orbit
- True vortexing action
The Multi-MicroPlate Genie® accepts most SBS microplate types, including 384-well plates. The mixer's high speed and small orbit combine to offer true vortexing action in each well of the microplate for complete and uniform mixing. A built-in timer operates continuously or from 1 to 60 minutes for analog model, and to 99 minutes for digital model. Digital model incorporates pulsing feature.
Certifications: CE marked.
BodyTextAccessories: An accessory tray to increase capacity up to 8 microplates is available.
Ordering information: The 230 V model is supplied without a plug.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89202-334) is also available, please see separate product page.
Customers Who Bought This Also Bought