Hemostats for Soldering, Aven Tools
Fournisseur: Aven Tools
Certificats
A propos de cet article
Versatile tools for use as clamps, heat sinks, third hands, and more while soldering
With three-position locking device. Stainless steel construction.
Customers Who Bought This Also Bought