Scoop, Stainless Steel, Polar Ware
# de catalogue 56921-006
Fournisseur: Polar Ware
Spécifications
- Capacity:1183 ml (40 oz.)
- Cat. no.:56921-006
- Bowl Dimensions (W×L×H):12.7ia.×19 cm (5×7½")
- Overall Length:30.5 cm (12")
- Supplier No.:T7010
Spécifications
A propos de cet article
Heavy-duty scoop has seamless body and strong, tubular handle for easy handling and cleaning.
Customers Who Bought This Also Bought