VWR® Rubber Acid Bucket
# de catalogue 17302-117
Fournisseur: VWR International
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Capacity:11.3 L (3 gal.)
- Color:Black
Spécifications
A propos de cet article
Resists the corrosive action of a wide variety of chemicals.
- Great chemical resistance
- Safety grip construction
- Stainless steel handle with rubber grip
- Internal graduated markings
Features a stainless steel bail-type handle with hard rubber grip, and safety grip construction at bottom for steady pouring. With graduated markings on inside: 1–12 quarts and 1–3 gallons.
Customers Who Bought This Also Bought