Technik™ Stainless Steel Pliers, Aven
Fournisseur: Aven Tools
Certificats
A propos de cet article
Pliers feature dual leaf springs and box joint construction for comfortable action.
- Made of mirror polished stainless steel
- Induction hardened cutting edges
- ESD-safe grips
Pliers feature dual leaf springs and box joint construction for comfortable action.
Customers Who Bought This Also Bought