ContactUsLinkComponent
IKA® Works Reciprocating Shakers, 230 V, HS 260
IKA® Works Reciprocating Shakers, 230 V, HS 260
Fournisseur:  IKA Works
Certificats
IKA® Works Reciprocating Shakers, 230 V, HS 260
IKA® Works Reciprocating Shakers, 230 V, HS 260
Fournisseur:  IKA Works
Certificats
Spécifications
  • Catalog No:
  • 33998-366
  • 33998-370
  • Dimensions:
  • 36W x 42D x 10H cm (143/16 x 1617/32 x 315/16")
  • 36W x 42D x 10H cm (143/16 x 1617/32 x 315/16")
  • Max. load:
  • 7.5 kg (16.5 lbs.)
  • 7.5 kg (16.5 lbs.)
  • Operating temperature range:
  • 5 to 50°C (41 to 122°F)
  • 5 to 50°C (41 to 122°F)