ContactUsLinkComponent
Four-Edged Glass Scoring Knife, Wale Apparatus
Four-Edged Glass Scoring Knife, Wale Apparatus
# de catalogue 32861-011
Fournisseur:  Wale Apparatus
Four-Edged Glass Scoring Knife
Four-Edged Glass Scoring Knife, Wale Apparatus
# de catalogue 32861-011
Fournisseur:  Wale Apparatus
Spécifications
  • Description:
    Glass Scoring Knife
  • Cat. no.:
    32861-011
  • Supplier No.:
    90-6050
Spécifications
A propos de cet article
The four cutting edges of this knife ensures a greater ease of use and longer life and it is ideally designed for cutting glass and quartz tubing and rods.
  • Retractable and replaceable blade
Unique square collet holds high-quality tungsten-carbide steel blade. User can expose as much of blade as desired and easily replace a worn blade.
Ordering information: Knife comes complete with four-edged blade. Replacement blades available.
Customers Who Bought This Also Bought